Anti-BPI Rabbit Polyclonal Antibody

GTX105150
89354-134EA 258.54 CAD
89354-134
Anti-BPI Rabbit Polyclonal Antibody
This gene encodes a lipopolysaccharide binding protein. It is associated with human neutrophil granules and has bactericidal activity on gram-negative organisms. [provided by RefSeq]

Type: Primary
Antigen: BPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR