Anti-BRDT Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX100201
89348-278EA 1222.84 CAD
89348-278
Anti-BRDT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to BRDT (bromodomain, testis-specific)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BRDT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR