Anti-BrdU Mouse Monoclonal Antibody

Supplier: GeneTex
GTX27781
89352-296EA 565.86 CAD
89352-296
Anti-BrdU Mouse Monoclonal Antibody
Antibodies
The immunocytochemical detection of bromodeoxyuridine (BrdU) incorporated into DNA is a powerful tool to study the cytokinetics of normal and neoplastic cells. In vitro or in vivo labeling of tumor cells with the thymidine analogue BrdU and the subsequent detection of incorporated BrdU with specific anti-BrdU monoclonal antibodies is an accurate and comprehensive method to quantitate the degree of DNA-synthesis. BrdU is incorporated into the newly synthezised DNA of the S-phase cells and can thus provide an estimate for the fraction of cells in S-phase. Also dynamic proliferative information (such as the S-phase transit rate and the potential doubling time) can be obtained, by means of bivariate BrdU/DNA flow cytometric analysis.

Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR