Anti-PTK6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107671
89355-052EA 881.1 CAD
89355-052
Anti-PTK6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Brk (Brk protein tyrosine kinase 6)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PTK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR