Anti-BSA Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX79816
89366-648EA 727.95 CAD
89366-648
Anti-BSA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to BSA (Biotin)

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: BSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR