Anti-BSG Mouse Monoclonal Antibody [clone: 9H5]

Supplier: GeneTex
GTX84801
89366-802EA 1067.2 CAD
89366-802
Anti-BSG Mouse Monoclonal Antibody [clone: 9H5]
Antibodies
Mouse monoclonal antibody [9H5] to BSG

Recommended Starting Dilutions: WB 1:1500~3000 IHC 1:50 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: 9H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR