Anti-BTNL2 Rabbit Polyclonal Antibody

GTX110026
89356-148EA 258.54 CAD
89356-148
Anti-BTNL2 Rabbit Polyclonal Antibody
Negative regulator of T cell proliferation.

Type: Primary
Antigen: BTNL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR