Anti-BTN3A2 Mouse Monoclonal Antibody [clone: 1A6]

Supplier: GeneTex

GTX84773
89366-800EA 970.77 CAD
89366-800
Anti-BTN3A2 Mouse Monoclonal Antibody [clone: 1A6]
Antibodies
Mouse monoclonal antibody [1A6] to BTN3A2

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BTN3A2
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR