Anti-BUB3 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX113595
89357-636EA 623.96 CAD
89357-636
Anti-BUB3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to BUB3 (budding uninhibited by benzimidazoles 3 homolog (yeast))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BUB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR