Anti-CRP Mouse Monoclonal Antibody [clone: C3]

Supplier: GeneTex

GTX10027
89350-586EA 428.64 CAD
89350-586
Anti-CRP Mouse Monoclonal Antibody [clone: C3]
Antibodies
Mouse monoclonal [C3] to C Reactive Protein

AP: Use at an assay dependent dilution. ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CRP
Clonality: Monoclonal
Clone: C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR