Anti-CRP Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX101262
89348-936EA 623.96 CAD
89348-936
Anti-CRP Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to C-reactive protein (C-reactive protein, pentraxin-related)

Suggested starting dilutions are as follows: WB: 1:500-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR