Anti-C1Q Sheep Polyclonal Antibody

Supplier: GeneTex

GTX28779
89352-528EA 802.59 CAD
89352-528
Anti-C1Q Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal to C1q

RID and Rocket IEP: 0.1?L antiserum/cm2 in gel vs 20?L fresh plasma, NT - 1/2Double Diffusion: 10?L antiserum vs 20?L fresh plasmaSecond Antibody: 100?L antiserum vs 10?L fresh plasma The use of 3% PEG 6000 with 1.2%agarose in a suitable buffer (such as TBE or Tris-barbital pH >8.2) is recommended. Results can be enhanced by washing completed agarose gels in saline, drying and finally stainingwith a protein stain such as coomassie blue. Suitability for use in immunohistochemical procedures, enzyme-linked immunosorbent assays and Western blot has not been assessed butuse in such assays should not necessarily be excluded.

Type: Primary
Antigen: C1Q
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR