Anti-C3 Goat Polyclonal Antibody

Supplier: GeneTex

GTX40423
89363-184EA 565.64 CAD
89363-184
Anti-C3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to C3 (FITC)

IF: 1/15 - 1/50. Acetone fixation of the antigen source is recommended prior to staining. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR