Anti-Le-A Mouse Monoclonal Antibody

GTX75539
89367-890EA 570.96 CAD
89367-890
Anti-Le-A Mouse Monoclonal Antibody
CA19-9 is a carbohydrate antigenic determinant identified as a sialylated lacto-N-fucopentose II, an oligosaccharide biochemically related to the Lewis A blood group antigen. In malignant tissues positive staining of tumour cells is observed in most adenocarcinomas of the pancreas and stomach, and in a large proportion of colon and gall bladder tumours. The antigen is also expressed in approximately half of primary and metastatic ovarian tumours. In normal tissues positive staining is also observed in columnor epithelium of the pancreas stomach, liver, gall bladder and in bronchial glands of the lung.

Type: Primary
Antigen: Le-A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR