Anti-CALD1 Mouse Monoclonal Antibody [clone: C21]

Supplier: GeneTex
GTX11016
89356-204EA 565.19 CAD
89356-204
Anti-CALD1 Mouse Monoclonal Antibody [clone: C21]
Antibodies
Mouse monoclonal [C21] to Caldesmon

DID: Use at an assay dependent dilution. ELISA: Use at an assay dependent dilution. The minimum antibody titer of 1/2500 was determined in an ELISA with turkey gizzard or pig stomach caldesmon coated at 10 g/ml on polyvinyl microtiter plates. IF: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Caldesmon occurs in both a high molecular weight (120-150 KD) and a low molecular weight (60-80KD) form, depending on the tissue in which it is located. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CALD1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Gerbil
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR