Anti-CAM Rabbit Monoclonal Antibody

GTX61343
89363-946EA 556.26 CAD
89363-946
Anti-CAM Rabbit Monoclonal Antibody
Calmoduin (CaM) is a calcium modulator protein and a transducer of calcium signals . Upon calcium binding, calmodulin undergoes conformational changes and binds and modulates a diverse array of proteins. Calcium-bound CaM (Ca2+-CaM) can assume a variety of shapes depending on the target . Ca2+-CaM binds many kinases, phosphatases, signaling proteins, and structural proteins affecting a wide variety of processes including neurotransmitter release, muscle contraction, metabolism, apoptosis, inflammation, membrane protein organization, and cytoskeleton movement (2, 4-5).

Type: Primary
Antigen: CAM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR