Anti-CAMKI Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX15810
89360-130EA 624.75 CAD
89360-130
Anti-CAMKI Rabbit Polyclonal Antibody
Antibodies
Calcium / calmodulin-dependent kinases (CaMKs) are a subfamily of the serine/threonine protein kinase family that includes CaMKI, CaMKII, and CaMKIV. There have been identified multiple substrates for CaMKI, including synapsin 1, synapsin 2, CREB, and CFTR, and its activation is dependent upon phosphorylation of Thr 177 by CaMK kinase (CaMKK). Studies suggest that CaMKI plays an important role in retinal function and its expression is regulated by light stimulation.

  • IP: 1/200. WB: 1/500. Detects a band of approximately 36-38 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CAMKI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR