Anti-Candida Albicans Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX40005
89363-068EA 662.16 CAD
89363-068
Anti-Candida Albicans Rabbit Polyclonal Antibody
Antibodies
Candida albicans is the most frequently isolated fungal pathogen of humans, affecting immuocompromised patients ranging from premature infants to AIDS sufferers. Systemic infections have an attributed mortality of 30-50%. C. albicans is a diploid organism which has eight sets of homologous chromosomes. Its genome size is about 16 Mb (haploid), about 30% greater than S. cerevisiae (baker?s yeast).

Type: Primary
Antigen: Candida Albicans
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Fungi
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR