Anti-Canine Distemper Virus Mouse Monoclonal Antibody

Supplier: GeneTex

GTX28353
89352-476EA 428.64 CAD
89352-476
Anti-Canine Distemper Virus Mouse Monoclonal Antibody
Antibodies
This antibody recognizes canine Distemper Virus

Type: Primary
Antigen: Canine Distemper Virus
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR