Anti-CNR2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX78039
89366-486EA 728.03 CAD
89366-486
Anti-CNR2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Cannabinoid Receptor 2 (CB2)

Type: Primary
Antigen: CNR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR