Anti-CAPZB Goat Polyclonal Antibody

Supplier: GeneTex
GTX74496
89365-458EA 593.52 CAD
89365-458
Anti-CAPZB Goat Polyclonal Antibody
Antibodies
Goat polyclonal to CAPZB

For WB: Use at a concentration of 0.5 - 2.0 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CAPZB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR