Anti-CPM Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX106203
89354-540EA 623.96 CAD
89354-540
Anti-CPM Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Carboxypeptidase M (carboxypeptidase M)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CPM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR