Anti-Carboxypeptidase Y Rabbit Polyclonal Antibody

Supplier: Genetex

GTX40574
89363-238EA 549.41 CAD
89363-238
Anti-Carboxypeptidase Y Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Carboxypeptidase Y
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR