Anti-TNNI3 Mouse Monoclonal Antibody [clone: 10B11]

Supplier: GeneTex
GTX10225
89350-718EA 512.31 CAD
89350-718
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 10B11]
Antibodies
Mouse monoclonal [10B11] to cardiac Troponin I

Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: 10B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR