Anti-TNNI3 Mouse Monoclonal Antibody [clone: 2D5]

Supplier: GeneTex
GTX72420
89353-026EA 525.41 CAD
89353-026
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 2D5]
Antibodies
Mouse monoclonal [2D5] to Cardiac Troponin I

Immunohistochemistry 1:25-1:50.( For formalin-fixed paraffin-embedded (FFPE) tissue sections antigen demasker (10 mM citrate buffer pH 6.0 ) is requied. Boil the tissue 10-20 mins in this buffer, followed by cooling to RT. This antibody can be used on formalin-fixed, paraffin-embedded tissue sections. Prolonged fixation in buffered formalin can destroy the epitope. The antibody may be used at a dilution of 1:25-1:50 with LAB/Probe. When using paraffin-embedded and formalin-fixed tissues, high temperature antigenic unmasking technique (see instructions below) is strongly recommended for consistent and reproducible results. This product can also be used on frozen tissue sections. The optimal conditions should be determined by the individual laboratory.

Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: 2D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR