Anti-TNNI3 Mouse Monoclonal Antibody [clone: 8E10]

Supplier: GeneTex

GTX10232
89350-726EA 565.64 CAD
89350-726
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 8E10]
Antibodies
Mouse monoclonal [8E10] to cardiac Troponin I

Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: 8E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR