Anti-TNNI3 Mouse Monoclonal Antibody [clone: M46]

Supplier: GeneTex

GTX10238
89350-730EA 565.64 CAD
89350-730
Anti-TNNI3 Mouse Monoclonal Antibody [clone: M46]
Antibodies
Mouse monoclonal [M46] to cardiac Troponin I

Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: M46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR