Anti-Cardiac Troponin T Mouse Monoclonal Antibody [clone: 3D6]

GTX10218
89350-712EA 443.36 CAD
89350-712
Anti-Cardiac Troponin T Mouse Monoclonal Antibody [clone: 3D6]
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.

Type: Primary
Antigen: Cardiac Troponin T
Clonality: Monoclonal
Clone: [3D6]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR