Anti-TNNT2 Mouse Monoclonal Antibody [clone: 7A9]

Supplier: GeneTex

GTX10221
89350-714EA 504.24 CAD
89350-714
Anti-TNNT2 Mouse Monoclonal Antibody [clone: 7A9]
Antibodies
Mouse monoclonal [7A9] to cardiac Troponin T

AP: Use at an assay dependent dilution. ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 35.4 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNNT2
Clonality: Monoclonal
Clone: 7A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog, Goat, Rabbit, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR