Anti-Casein Kinase 1 D Goat Polyclonal Antibody

Supplier: GeneTex

GTX10877
89355-692EA 624.4 CAD
89355-692
Anti-Casein Kinase 1 D Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Casein Kinase 1 delta

Recommended Starting Dilutions: Immunohistology - Paraffin: Max Dilution 1/200. Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR