Anti-CASP11 Rat Monoclonal Antibody

GTX10454
89350-380EA 488.93 CAD
89350-380
Anti-CASP11 Rat Monoclonal Antibody
Caspase 11 (also known as ICH3) gene encodes two protein products of 38 and 43 kDa that belong to the Ced3/ICE family of cystein proteases. Caspase 11 is an upstream caspase with a long prodomain. It undergoes a cleavage upon activation, leading to p20 and p10 subunits. Monoclonal antibodies reacting specifically with caspase 11 are useful tool for the study of the protease network, involved in development and regulation, governing the life and death of cells and tissues.

Type: Primary
Antigen: CASP11
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR