Anti-CASP12 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX28117
89362-726EA 749.03 CAD
89362-726
Anti-CASP12 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Caspase 12

For WB: Use at a dilution of 1:500-1:1,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CASP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR