Anti-CASP3 Mouse Monoclonal Antibody [clone: C33]

Supplier: GeneTex

GTX59551
89363-712EA 579.54 CAD
89363-712
Anti-CASP3 Mouse Monoclonal Antibody [clone: C33]
Antibodies
Mouse monoclonal antibody [C33] to Caspase 3

Western blotting (0.5-4 µg/ml). The antibody recognizes both proform and the cleaved large fragment of caspase-3 in samples of human, mouse and rat origins. However, the optimal conditions should be determined individually.

Type: Primary
Antigen: CASP3
Clonality: Monoclonal
Clone: C33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR