Anti-CASP9 Mouse Monoclonal Antibody [clone: LAP6 96-2-22]

Supplier: GeneTex

GTX30245
89367-660EA 749.76 CAD
89367-660
Anti-CASP9 Mouse Monoclonal Antibody [clone: LAP6 96-2-22]
Antibodies
Mouse monoclonal to human Caspase 9

For WB: Use at a diluton of 1:500 - 1:1,000. Western blot with bands at 46~48 and 35 kDa representing the pro and active forms of Caspase 9, respectively. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CASP9
Clonality: Monoclonal
Clone: LAP6 96-2-22
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR