Anti-CTA1 Sheep Polyclonal Antibody

GTX28954
89367-080EA 765.15 CAD
89367-080
Anti-CTA1 Sheep Polyclonal Antibody
Catalase is the classical marker for peroxisomes and is the most abundant protein within peroxisomes.

Type: Primary
Antigen: CTA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR