Anti-CTA1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX26573
89361-944EA 453.84 CAD
89361-944
Anti-CTA1 Rabbit Polyclonal Antibody
Antibodies
Peroxidase-conjugated Rabbit Polyclonal antibody to Catalase

Recommended Starting Dilutions:WB: Use at 1:500- 1:2,000ELISA: Use at 1:10,000 - 1:50,000

Type: Primary
Antigen: CTA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR