Anti-CTA1 Rabbit Monoclonal Antibody

GTX61651
89364-106EA 556.26 CAD
89364-106
Anti-CTA1 Rabbit Monoclonal Antibody
Human catalase is an heme-containing peroxisomal enzyme that breaks down hydrogen peroxide to water and oxygen; it is implicated in ethanol metabolism, inflammation, apoptosis, aging and cancer. The 1. 5 A resolution human enzyme structure, both with and without bound NADPH, establishes the conserved features of mammalian catalase fold and assembly, implicates Tyr370 as the tyrosine radical, suggests the structural basis for redox-sensitive binding of cognate mRNA via the catalase NADPH binding site, and identifies an unexpectedly substantial number of water-mediated domain contacts . Because catalase has a low affinity for H2O2, it has been suggested that glutathione peroxidase clears most H2O2 within the erythrocyte and that catalase is of little import. It has been hypothesized that erythrocyte catalase might function to protect heterologous somatic cells against challenge by high levels of exogenous H2O2 (e.g., in areas of inflammation) .

Type: Primary
Antigen: CTA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR