Anti-CTSB Mouse Monoclonal Antibody

GTX46395
89363-604EA 472.59 CAD
89363-604
Anti-CTSB Mouse Monoclonal Antibody
Mouse monoclonal antibody [CB131] to Cathepsin B

Recommended Starting Dilutions: Immunohistology - Paraffin: 1/40 - 1/80 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CTSB
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR