Anti-CTSD Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX20826
89351-552EA 623.94 CAD
89351-552
Anti-CTSD Rabbit Polyclonal Antibody
Antibodies
Cathepsin D has been reported to be involved in malignant tumor progression and with prognosis of various human cancers.

For IHC: This antibody may be diluted to a titer of 1:75-1:200 in an ABC method. Staining Protocol: We suggest an incubation period of 30-60 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions and antibody dilutions should be determined by the user. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CTSD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR