Anti-CAV1 Rabbit Monoclonal Antibody [clone: E249]

Supplier: GeneTex
GTX61090
89363-814EA 695.06 CAD
89363-814
Anti-CAV1 Rabbit Monoclonal Antibody [clone: E249]
Antibodies
Rabbit monoclonal antibody [E249] to Caveolin 1

Type: Primary
Antigen: CAV1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR