Anti-CAV1 Goat Polyclonal Antibody

Supplier: GeneTex

GTX89541
89366-996EA 675.53 CAD
89366-996
Anti-CAV1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Caveolin 1

Approx 21kDa and 25kDa observed in Human Heart, Lung and Spleen lysates. These bands correspond to earlier findings in literature with different antibodies (Wiechen et al Am J Pathol. 2001 Mar;158(3):833-9, PMID: 11238032 ). In transfected HEK293 transiently expressing Caveolin 1 a band of approx. 23kDa is observed. This band is not observed in the non-transfected HEK293. This protein has a calculated MW of 20.5kDa according to NP_001744.2. Recommended concentration: 0.3-1µg/ml. No cross-reactivity expected to Caveolin 2 and Caveolin 3

Type: Primary
Antigen: CAV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR