Anti-CCL20 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX45123
89363-572EA 998.26 CAD
89363-572
Anti-CCL20 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CCL20

For WB: Use at a dilution of 1:2,000. For IP: Use at a dilution of 1:500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CCL20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR