Anti-CCL3 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX81195
89366-714EA 689.8 CAD
89366-714
Anti-CCL3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MIP1 alpha

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR