Anti-CCR2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109684
89356-008EA 564.65 CAD
89356-008
Anti-CCR2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CCR2 (chemokine (C-C motif) receptor 2)

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR