Anti-CCRL2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX71119
89365-176EA 699.51 CAD
89365-176
Anti-CCRL2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to Human Chemokine ( C - C motif ) Receptor - like 2 ( CCRL2 )

Immunohistochemistry with formalin fixed paraffin embedded tissues. Not tested for other uses. IHC-P: Use at a dilution of 1/100 - 1/200.

Type: Primary
Antigen: CCRL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR