Anti-CD101 Mouse Monoclonal Antibody

Supplier: Genetex
GTX75986
89367-938EA 588.88 CAD
89367-938
Anti-CD101 Mouse Monoclonal Antibody
Mouse monoclonal antibody [BB27] to CD101

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD101
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR