Anti-ICAM2 Mouse Monoclonal Antibody [clone: B-T1]

Supplier: GeneTex

GTX74684
89365-530EA 796.53 CAD
89365-530
Anti-ICAM2 Mouse Monoclonal Antibody [clone: B-T1]
Antibodies
Mouse monoclonal antibody [B-T1] to ICAM2

Recommended Starting Dilutions: Flow Cytometry: 1/20 - 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ICAM2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR