Anti-ITGB4 Mouse Monoclonal Antibody [clone: 450-9D]

Supplier: GeneTex

GTX75043
89365-676EA 916.37 CAD
89365-676
Anti-ITGB4 Mouse Monoclonal Antibody [clone: 450-9D]
Antibodies
Mouse monoclonal antibody [450-9D] to Integrin beta 4

Recommended Starting Dilutions: Flow Cytometry: 1/25 - 1/100 Immunohistology - Frozen: 1/500 - 1/1000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ITGB4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR