Anti-ENG Mouse Monoclonal Antibody

GTX73783
89353-276EA 667.26 CAD
89353-276
Anti-ENG Mouse Monoclonal Antibody
This antibody reacts with 95 kD (monomer) and 190 kD (dimer) proteins. CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF-b1 and b3 with high affinity but not to TGF-b2.

Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR