Anti-VCAM1 Mouse Monoclonal Antibody

Supplier: Genetex
GTX76604
89366-152EA 615.88 CAD
89366-152
Anti-VCAM1 Mouse Monoclonal Antibody
Recognises human VCAM-1, a 110kD molecule whose ligand is VLA4. The antigen is expressed on activated endothelial cells and on some tissue macrophages, bone marrow fibroblasts and myoblasts.

Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR